Laboratorienotesbøger, Nalgene®
notesbog, grøn, Grid, A4, 96
115-0195
Polyethylene cover for durability in laboratory environments.
- Waterproof
- Grid or lined
- Place for signature on the bottom of every page
96 pages.
- Papirformat:A4
- Model:Grid
- Farve:grøn
- Beskrivelse:notesbog
- Antal af sider:96
- Cat. No.:115-0195
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning