VWR®, Block Weights, Iron, Class M2
10 kg, 1600 ± mg
611-8388
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
: Conforms to OIML, COFRAC accredited.
- Vægt:10 kg
- Tolerance:1600 ± mg