Orders LinkContactUsLinkComponent
VWR®, Block Weights, Iron, Class M2
10 kg, 1600  ± mg
  611-8388
VWR®, Block Weights, Iron, Class M2
  611-8388
VWR®, Block Weights, Iron, Class M2
VWR®, Block Weights, Iron, Class M2
10 kg, 1600  ± mg
1.250,00 kr
1 * 1 Items
Rectangular weights made from cast iron, permanently magnetised.
  • Indelible unique identification number guarantees traceability
  • Painted grey
  • Adjustment cavity sealed with a lead plug
 : Conforms to OIML, COFRAC accredited.
  • Vægt:
    10 kg
  • Tolerance:
    1600  ± mg