VWR®, Block Weights, Iron, Class M2
50 kg, 7500 ± mg
611-8386
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
Documents
- Vægt:50 kg
- Seq:08
- Tolerance:7500 ± mg