Orders LinkContactUsLinkComponent
VWR®, Block Weights, Iron, Class M2
VWR®, Block Weights, Iron, Class M2
20 kg, 3000 ± mg
  611-8385
VWR®, Block Weights, Iron, Class M2
  611-8385
VWR®, Block Weights, Iron, Class M2
VWR®, Block Weights, Iron, Class M2
20 kg, 3000 ± mg
1.290,00 kr
1 * 1 Items
Rectangular weights made from cast iron, permanently magnetised.
  • Indelible unique identification number guarantees traceability
  • Painted grey
  • Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
  • Vægt:
    20 kg
  • Seq:
    07
  • Tolerance:
    3000 ± mg