VWR®, Block Weights, Iron, Class M2
20 kg, 3000 ± mg
611-8385
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
Documents
- Vægt:20 kg
- Seq:07
- Tolerance:3000 ± mg