VWR®, Block Weights, Iron, Class M2
10 kg, 1600 ± mg
Katalog # 611-8384
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
Documents
Specifikationer
- Vægt:10 kg
- Seq:06
- Tolerance:1600 ± mg