VWR®, Block Weights, Iron, Class M2
5 kg, 750 ± mg
Katalog # 611-8383
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Rectangular weights made from cast iron, permanently magnetised.
- Indelible unique identification number guarantees traceability
- Painted grey
- Adjustment cavity sealed with a lead plug
Certifications: Conforms to OIML, COFRAC accredited.
Documents
Specifikationer
- Vægt:5 kg
- Seq:05
- Tolerance:750 ± mg