VWR®, Calibration Weights, Class M2
2 kg, 300 ± mg
611-8380
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
- Vægt:2 kg
- Seq:14
- Tolerance:300 ± mg