VWR®, Calibration Weights, Class M2
1 kg, 160 ± mg
Catalog # 611-8379
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
Specifications
- Vægt:1 kg
- Seq:13
- Tolerance:160 ± mg