VWR®, Calibration Weights, Class M2
500 g, 75 ± mg
Katalog # 611-8378
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Leveringsinformation: Supplied in a plastic box.
Documents
Specifikationer
- Vægt:500 g
- Seq:12
- Tolerance:75 ± mg