Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
500 g, 75 ± mg
Katalog # 611-8378
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M2
Katalog # 611-8378
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
500 g, 75 ± mg
Price
933,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Test weights in either stainless steel or aluminium.
Leveringsinformation: Supplied in a plastic box.
Specifikationer
  • Vægt:
    500 g
  • Seq:
    12
  • Tolerance:
    75 ± mg