VWR®, Calibration Weights, Class M2
200 g, 30 ± mg
Katalog # 611-8377
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Documents
Specifikationer
- Vægt:200 g
- Seq:11
- Tolerance:30 ± mg