Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
5 g, 5 ± mg
Katalog # 611-8372
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M2
Katalog # 611-8372
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
5 g, 5 ± mg
Price
384,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Specifikationer
  • Vægt:
    5 g
  • Seq:
    06
  • Tolerance:
    5 ± mg