VWR®, Calibration Weights, Class M2
100 g, 16 ± mg
Katalog # 611-8359
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Leveringsinformation: Supplied in a plastic box.
Documents
Specifikationer
- Vægt:100 g
- Seq:26
- Tolerance:16 ± mg