VWR®, Calibration Weights, Class M2
20 g, 8 ± mg
Katalog # 611-8357
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Leveringsinformation: Supplied in a plastic box.
Documents
Specifikationer
- Vægt:20 g
- Seq:24
- Tolerance:8 ± mg