VWR®, Calibration Weights, Class M2
10 g, 6 ± mg
Katalog # 611-8356
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Test weights in either stainless steel or aluminium.
Leveringsinformation: Supplied in a plastic box.
Documents
Specifikationer
- Vægt:10 g
- Seq:23
- Tolerance:6 ± mg