VWR®, Calibration Weights, Class M1
10 kg, 500 ± mg
Katalog # 611-8297
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specifikationer
- Vægt:10 kg
- Seq:22
- Tolerance:500 ± mg