Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

1 kg, 50 ± mg
Katalog # 611-8294
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8294
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

1 kg, 50 ± mg

Price
1.240,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specifikationer
  • Vægt:
    1 kg
  • Seq:
    19
  • Tolerance:
    50 ± mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.