VWR®, Calibration Weights, Class M1
500 g, 25 ± mg
611-8289
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
- Vægt:500 g
- Seq:18
- Tolerance:25 ± mg
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.