Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Calibration Weights, Class M1

200 g, 10 ± mg
Katalog # 611-8288
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8288
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

200 g, 10 ± mg

Price
518,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

 : All weights are supplied in plastic cases.

Specifikationer
  • Vægt:
    200 g
  • Seq:
    17
  • Tolerance:
    10 ± mg
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.