VWR®, Calibration Weights, Class M1
200 mg, 0,6 ± mg
Katalog # 611-8279
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifikationer
- Vægt:200 mg
- Seq:08
- Tolerance:0,6 ± mg