VWR®, Calibration Weights, Class M1
100 mg, 0,5 ± mg
Catalog # 611-8278
Supplier: VWR Collection
Jump to:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specifications
- Vægt:100 mg
- Seq:07
- Tolerance:0,5 ± mg