VWR®, Calibration Weights, Class M1
20 mg, 0,3 ± mg
611-8276
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
- Vægt:20 mg
- Seq:05
- Tolerance:0,3 ± mg
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.