Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1
10 mg, 0,25 ± mg
Katalog # 611-8275
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8275
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
10 mg, 0,25 ± mg
Price
359,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
 : All weights are supplied in plastic cases.
Specifikationer
  • Vægt:
    10 mg
  • Seq:
    04
  • Tolerance:
    0,25 ± mg