VWR®, Calibration Weights, Class M1
10 mg, 0,25 ± mg
Katalog # 611-8275
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifikationer
- Vægt:10 mg
- Seq:04
- Tolerance:0,25 ± mg