VWR®, Calibration Weights, Class M1
10 kg, 500 ± mg
611-8271
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
- Vægt:10 kg
- Seq:47
- Tolerance:500 ± mg
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.