Skilletragte, Dråbeformet, Nalgene®
125 ml, 28 mm
Katalog # 527-0005
Leverandør: Thermo Fisher Scientific
Skilletragte, Dråbeformet, Nalgene®
Katalog # 527-0005
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
PPCO translucent, TFE stopcock, PP screw closure.
- Leakproof closure and stopcock
- Very good chemical resistance
- Autoclavable except for stopcock assembly
- TFE stopcock requires no lubrication
Specifikationer
- Kapacitet:125 ml
- Ø hætte:28 mm
- Cat. No.:527-0005
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning