VWR®, Calibration Weights, Class M1
500 g, 25 ± mg
Katalog # 611-8267
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifikationer
- Vægt:500 g
- Seq:43
- Tolerance:25 ± mg



