Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

100 g, 5 ± mg
Catalog # 611-8265
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalog # 611-8265
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

100 g, 5 ± mg

Price
219,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specifications
  • Vægt:
    100 g
  • Seq:
    41
  • Tolerance:
    5 ± mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.