VWR®, Calibration Weights, Class M1
50 g, 3,0 ± mg
Katalog # 611-8264
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
Specifikationer
- Vægt:50 g
- Seq:40
- Tolerance:3,0 ± mg