Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Calibration Weights, Class M1

10 g, 2,0 ± mg
Katalog # 611-8262
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8262
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

10 g, 2,0 ± mg

Price
155,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

 : All weights are supplied in plastic cases.

Specifikationer
  • Vægt:
    10 g
  • Seq:
    38
  • Tolerance:
    2,0 ± mg