VWR®, Calibration Weights, Class M1
5 g, 1,6 ± mg
611-8261
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
- Vægt:5 g
- Seq:37
- Tolerance:1,6 ± mg
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.