Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Calibration Weights, Class M1

2 g, 1,2 ± mg
Katalog # 611-8260
Leverandør:   VWR Collection
VWR®, Calibration Weights, Class M1
Katalog # 611-8260
Leverandør:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

2 g, 1,2 ± mg

Price
132,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

 : All weights are supplied in plastic cases.

Specifikationer
  • Vægt:
    2 g
  • Seq:
    36
  • Tolerance:
    1,2 ± mg