VWR®, Calibration Weights, Class M1
100 mg, 0,5 ± mg
Katalog # 611-8256
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Leveringsinformation: All weights are supplied in plastic cases.
Documents
Specifikationer
- Vægt:100 mg
- Seq:31
- Tolerance:0,5 ± mg