Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pulvertragt, Nalgene®

Pulvertragt, Nalgene®

64 mm, 17 mm, PP, 22 mm, Powder funnels, 68 ml, 67 mm hieght
Katalog # 221-0090
   Lav fremstillingspåvirkning
Leverandør:   Thermo Fisher Scientific
Pulvertragt, Nalgene®
Katalog # 221-0090
   Lav fremstillingspåvirkning
Pulvertragt, Nalgene®
Pulvertragt, Nalgene®

64 mm, 17 mm, PP, 22 mm, Powder funnels, 68 ml, 67 mm hieght

Price
534,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents

PPCO, gennemskinnelig.

  • Meget god kemisk resistens
  • De ydre ribber giver en bedre luftudtømning
  • Autoklaverbare

Til overførsel af pulveragtige materialer. Den cylindrisk udformede stilk forhindrer blokeringer.

This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.

Specifikationer
  • Tragt udv. Ø:
    64 mm
  • Beskrivelse:
    Powder funnels, 68 ml, 67 mm hieght
  • Materiale:
    PP
  • Seq:
    01
  • Stilk udv. Ø:
    17 mm
  • Stilklængde:
    22 mm
  • Cat. No.:
    221-0090
  • Miljøcertificering:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.