VWR®, Calibration Weights, Class M1
20 mg, 0,3 ± mg
Katalog # 611-8254
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specifikationer
- Vægt:20 mg
- Seq:29
- Tolerance:0,3 ± mg