Dunke, engangsbrug, Nalgene®
Single-use HDPE carboy, sterile, 20 L, Carboy, HDPE, hvid, 284 mm, 64 mm, 502 mm, 83B
HDPE, gennemskinnelig med hvid PP-lukning.
- Good chemical resistance
- Heat resistant from –100 to +120 °C
- Complies with the requirements of USP class VI, non cytotoxic
Mere sikkert end glas. Ideel til opbevaring af sterile væsker og farmaceutiske/biotekniske produkter. Den ekstra brede munding gør fyldning og tømning lettere.
Eliminate cleaning, prep and set-up costs with ready-to-use Nalgene™ single-use HDPE carboys. Supplied sterile for one-time use, the high-density polyethelyne vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
- Udv.Ø:284 mm
- Gevind:83B
- Model:Carboy
- Kapacitet:20 L
- Farve:hvid
- Beskrivelse:Single-use HDPE carboy, sterile
- Højde:502 mm
- Materiale:HDPE
- Seq:01
- Hals indv. Ø:64 mm
- Cat. No.:215-1182
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning