Ekssikatoelåg, DURAN®
Med NS 24/29, NOVUS, 200 DN
Katalog # 467-1280
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear plain lids for desiccators. Lids have either knob or NS-connection for stop cork (Novus) or thread connection for stop cork (Mobilex).
: According to DIN ISO 13130
Specifikationer
- Nominel størrelse:200 DN
- Model:Med NS 24/29, NOVUS
- Seq:012