Ekssikatoelåg, DURAN®
Med knop, 250 DN
Catalog # 467-1185
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear plain lids for desiccators. Lids have either knob or NS-connection for stop cork (Novus) or thread connection for stop cork (Mobilex).
: According to DIN ISO 13130
Specifications
- Nominel størrelse:250 DN
- Model:Med knop
- Seq:006