Orders LinkContactUsLinkComponent
Ekssikatoelåg, DURAN®
Med knop, 250 DN
Catalog # 467-1185
Supplier:   DWK Life Sciences
Ekssikatoelåg, DURAN®
Catalog # 467-1185
Supplier:  DWK Life Sciences
Ekssikatoelåg, DURAN®
Ekssikatoelåg, DURAN®
Med knop, 250 DN
Price
1.070,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear plain lids for desiccators. Lids have either knob or NS-connection for stop cork (Novus) or thread connection for stop cork (Mobilex).
 : According to DIN ISO 13130
Specifications
  • Nominel størrelse:
    250 DN
  • Model:
    Med knop
  • Seq:
    006