VWR®, Flaskedispenser
Product Details & Documents
Variabel volumen, autoklaverbar.
- Valve system with steel spring
- PP with precision glass cylinder
- Quick and easy to use volume setting system
- Additional rotating fine volume adjustment for right or left handed use
- Adapter allows rotation of the dispenser on bottles to the desired position
These standard PP units, with glass cylinder, are suitable for aqueous liquids, dilute acids and alkalis - not for use with solvents and hydrofluoric acid which attack PP (for bottles with GL 32 thread). The glass cylinder and PTFE-encased glass piston, with PP body, models are designed for aqueous solutions, weakly concentrated acids, alkaline solutions and crystallising liquids compatible with PP. The amber glass cylinders, clear glass piston, with PP body, are suitable for light-sensitive liquids, compatible with PP.
Leveringsinformation: Supplied with two adapters (GL 40 and GL 45) and calibration certificate.
Specifikationer
- Volumen:10 - 50 ml
- Inddeling:1,00 ml
Accessories
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Spare piston
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Valve block unit
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Intake tube
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Plunger PTFE
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Plunger PTFE
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Brown glass cylinder
Produktinformation
- Katalog #VolumenTilgængelighedPrice
- Brown glass cylinder