Orders LinkContactUsLinkComponent
VWR®, Flaskedispenser
VWR®, Flaskedispenser
2 - 10 ml, 0,50 ml
Catalog # 612-4750
Supplier:   VWR Collection
VWR®, Flaskedispenser
Catalog # 612-4750
Supplier:  VWR Collection
VWR®, Flaskedispenser
VWR®, Flaskedispenser
2 - 10 ml, 0,50 ml
Price
2.600,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Variabel volumen, autoklaverbar.
  • Valve system with steel spring
  • PP with precision glass cylinder
  • Quick and easy to use volume setting system
  • Additional rotating fine volume adjustment for right or left handed use
  • Adapter allows rotation of the dispenser on bottles to the desired position
These standard PP units, with glass cylinder, are suitable for aqueous liquids, dilute acids and alkalis - not for use with solvents and hydrofluoric acid which attack PP (for bottles with GL 32 thread). The glass cylinder and PTFE-encased glass piston, with PP body, models are designed for aqueous solutions, weakly concentrated acids, alkaline solutions and crystallising liquids compatible with PP. The amber glass cylinders, clear glass piston, with PP body, are suitable for light-sensitive liquids, compatible with PP.
Delivery information: Supplied with two adapters (GL 40 and GL 45) and calibration certificate.
Specifications
  • Volumen:
    2 - 10 ml
  • Inddeling:
    0,50 ml
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-6682
    Spare piston
    10 ml
    Log in for availability
    1 * 1 Items - 896,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-6684
    Valve block unit
    50 ml
    Log in for availability
    1 * 1 Items - 1.370,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-6686
    Intake tube
    2 - 10 ml and 10 - 50 ml
    Log in for availability
    1 * 2 Items - 689,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-3673
    Plunger PTFE
    10 ml
    Log in for availability
    1 * 1 Items - 1.340,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-3674
    Plunger PTFE
    50 ml
    Log in for availability
    1 * 1 Items - 1.530,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-3675
    Brown glass cylinder
    10 ml
    Log in for availability
    1 * 1 Items - 1.340,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Volumen
    Availability
    Price
  • 612-3676
    Brown glass cylinder
    50 ml
    Log in for availability
    1 * 1 Items - 1.530,00 kr