Propper til separationstragte, DURAN®
500 ml
Katalog # SCOT210134404
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Made of DURAN®, borosilicate glass 3.3, clear.
- For glass manipulators
- Very good chemical resistance
- Very good temperature resistance
Specifikationer
- Kapacitet:500 ml