Adaptere med standardslib, DURAN®
reduktion, 29/32 NS, 19/26 NS
Katalog # SCOT241142901
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
Certifications: DIN 12257
Specifikationer
- Konusstr.:29/32 NS
- Kappestr.:19/26 NS
- Model:reduktion