Orders LinkContactUsLinkComponent
Adaptere med standardslib, DURAN®
Adaptere med standardslib, DURAN®
reduktion, 24/29 NS, 14/23 NS
Katalog # SCOT241142301
Leverandør:   DWK Life Sciences
Adaptere med standardslib, DURAN®
Katalog # SCOT241142301
Leverandør:  DWK Life Sciences
Adaptere med standardslib, DURAN®
Adaptere med standardslib, DURAN®
reduktion, 24/29 NS, 14/23 NS
Price
222,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, reduction or expansion, with standard ground conical joints.
Certifications: DIN 12257
Specifikationer
  • Konusstr.:
    24/29 NS
  • Kappestr.:
    14/23 NS
  • Model:
    reduktion