Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Petriskåle, Steriplan®, DURAN®

Petriskåle, Steriplan®, DURAN®

Steriplan®, 180 mm, 30 mm
Katalog # 391-2870
Leverandør:   DWK Life Sciences
Petriskåle, Steriplan®, DURAN®
Katalog # 391-2870
Leverandør:  DWK Life Sciences
Petriskåle, Steriplan®, DURAN®
Petriskåle, Steriplan®, DURAN®

Steriplan®, 180 mm, 30 mm

Price
96,10 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Soda-lime glass. Hydrolytic class III. Diameter quoted in the table is the external diameter of the lid. The height refers to the height of just the base.

Documents

Specifikationer
  • Ø:
    180 mm
  • Version:
    Steriplan®
  • Højde:
    30 mm