Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Petriskåle, Steriplan®, DURAN®

Petriskåle, Steriplan®, DURAN®

Steriplan®, 100 mm, 15 mm
Catalog # 391-2830
Supplier:   DWK Life Sciences
Petriskåle, Steriplan®, DURAN®
Catalog # 391-2830
Supplier:  DWK Life Sciences
Petriskåle, Steriplan®, DURAN®
Petriskåle, Steriplan®, DURAN®

Steriplan®, 100 mm, 15 mm

Price
19,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Soda-lime glass. Hydrolytic class III. Diameter quoted in the table is the external diameter of the lid. The height refers to the height of just the base.

Documents

Specifications
  • Ø:
    100 mm
  • Version:
    Steriplan®
  • Højde:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.