Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør, DURAN®

Centrifugerør, DURAN®

Centrifugerør, rundbundet, 250 ml, 56×147 mm, uden cap
Katalog # 525-1114
Leverandør:   DWK Life Sciences
Centrifugerør, DURAN®
Katalog # 525-1114
Leverandør:  DWK Life Sciences
Centrifugerør, DURAN®
Centrifugerør, DURAN®

Centrifugerør, rundbundet, 250 ml, 56×147 mm, uden cap

Price
275,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Excellent chemical and thermal resistance
  • Max. RCF: 4000×g

Robust universal centrifuge tubes for a wide variety of applications.

525-1110 and 525-1114 not in accordance with DIN.

Certifications: DIN 58970 (Part 2) only for round bottom tubes.

Documents

Specifikationer
  • Lukkestørrelse:
    uden cap
  • Ø×L:
    56×147 mm
  • Kapacitet:
    250 ml
  • Beskrivelse:
    Centrifugerør, rundbundet
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.