Pipette adapters
Pipette adapters, red
Catalog # MARI5833001
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Adapters made of PS. Suitable for tubings for blood diluting pipettes and haemoglobin pipettes.
Not registered for in vitro diagnostic use within the EU.
Specifications
- Beskrivelse:Pipette adapters, red