Pipette adapters
Pipette adapters, white
MARI5833000
Adapters made of PS. Suitable for tubings for blood diluting pipettes and haemoglobin pipettes.
Not registered for in vitro diagnostic use within the EU.
- Beskrivelse:Pipette adapters, white