Centrifugerør
10 ml, 16/17×98 mm, uden cap, Conical bottom
Katalog # MARI3906001
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Borosilicate glass 3,3.
- Heavy walled
- Ungraduated
- Very good chemical resistance
- Very good temperature resistance
Short conical or round bottom.
Specifikationer
- Lukkestørrelse:uden cap
- Ø×L:16/17×98 mm
- Model:Conical bottom
- Kapacitet:10 ml
- Seq:01