Objektglas med fordybning
Objektglas med reaktionsbrønde, sort, 10, 8 mm
MARI1216550
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
- Ant. brønde:10
- Farve:sort
- Beskrivelse:Objektglas med reaktionsbrønde
- Seq:30
- Brønd Ø:8 mm