Urglas, DURAN®
Watch glass, DURAN® borosilicate glass, with fused edges, 60 mm
Catalog # 216-1114
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilikatglas 3.3, klar, med sammensmeltede kanter.
- Meget god kemisk resistens
- Høj temperaturresistens
- Minimal varmeudvidelse giver relativt høj resistens overfor temperaturændringer
DIN 12341
Certifications: DIN 12341
Specifications
- Ø:60 mm
- Beskrivelse:Watch glass, DURAN® borosilicate glass, with fused edges
- Seq:03