Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test Strips, Oil

Test Strips, Oil

QUANTOFIX® nitrite, 0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 0,5–80 - mg/L - NO₂⁻, Azo reaction with an aromatic amine, 40×40×120 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0312
Supplier:   MACHEREY-NAGEL
Test Strips, Oil
Catalog # 705-0312
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test Strips, Oil
Test Strips, Oil

QUANTOFIX® nitrite, 0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 0,5–80 - mg/L - NO₂⁻, Azo reaction with an aromatic amine, 40×40×120 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
335,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

This test paper allows the rapid and reliable detection of oil contamination of water and soil.

  • Colour changes: Blue to dark blue

For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample.

Specifications

Specifications

  • Kemisk metode:
    Azo reaction with an aromatic amine
  • Pakn:
    100 Tests
  • Beskrivelse:
    QUANTOFIX® nitrite
  • Dimensioner:
    40×40×120 mm (packaging)
  • Test pr. sæt:
    100 Tests
  • Område:
    0 • 1 • 5 • 10 • 20 • 40 • 80 - mg/L - NO₂⁻; 0,5–80 - mg/L - NO₂⁻
  • Indhold:
    100 test strips in a tube, instruction leaflet
  • Cat. No.:
    705-0312
  • Overensstemmelse:
    No hazardous material
  • Platform:
    Semi-quantitative test strips
  • Holdbarhed (måneder):
    30 (from production)
  • Vægt:
    44,3 g
  • Instrument:
    QUANTOFIX Relax
  • Parameter:
    All reagents included
  • Method:
    Nitrite - Griess reaction (white → red-violet)

Specifications